CAS RN 187389-52-2
CAS RN 187389-52-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Caspase Inhibitor I
Z-VAD-FMK, CAS 187389-52-2, is a cell-permeable, irreversible, pan-caspase inhibitor.
2
Novus Biologicals™ Z-VAD-FMK Inhibitor
Low molecular weight, organic compound that may help regulate biological process.