CAS RN 187397-18-8
IUPAC Name:
N-[5-(5-{2,4-dioxo-1,3,8-triazaspiro[4.5]decan-8-yl}pentanoyl)-2,4-dimethoxyphenyl]-4-(trifluoromethyl)benzene-1-sulfonamide hydrochloride
Synonyms:
N-[5-(5-{2,4-dioxo-1,3,8-triazaspiro[4.5]decan-8-yl}pentanoyl)-2,4-dimethoxyphenyl]-4-(trifluoromethyl)benzenesulfonamide hydrochloride
Molecular Weight (g/mol):
649.08
Molecular Formula:
C27H32ClF3N4O7S
InChi Key:
SZJZEJZLRDIHPB-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC(OC)=C(NS(=O)(=O)C2=CC=C(C=C2)C(F)(F)F)C=C1C(=O)CCCCN1CCC2(CC1)NC(=O)NC2=O
1
–
1
of
1
results
1
R&D Systems™ RS 102221 hydrochloride
CAS: 187397-18-8 Molecular Formula: C27H32ClF3N4O7S