CAS RN 18800-28-7
CAS RN 18800-28-7
IUPAC Name:
1-bromo-2-(2-bromoethoxy)benzene
Synonyms:
1-bromo-2-(2-bromoethoxy)benzene
Molecular Weight (g/mol):
279.96
Molecular Formula:
C8H8Br2O
InChi Key:
LOVVQTBEUIBASP-UHFFFAOYSA-N
SMILES:
BrCCOC1=CC=CC=C1Br
1
–
1
of
1
results
1
Thermo Scientific Chemicals 1-(2-Bromoethoxy)-2-bromobenzene, 95%, Thermo Scientific™
CAS: 18800-28-7 Molecular Formula: C8H8Br2O Molecular Weight (g/mol): 279.96 MDL Number: MFCD00634301 InChI Key: LOVVQTBEUIBASP-UHFFFAOYSA-N Synonym: 1-bromo-2-2-bromoethoxy benzene,1-2-bromoethoxy-2-bromobenzene,o-bromo-2-bromoethoxybenzene,benzene, 1-bromo-2-2-bromoethoxy PubChem CID: 2734928 IUPAC Name: 1-bromo-2-(2-bromoethoxy)benzene SMILES: BrCCOC1=CC=CC=C1Br