accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18800-30-1

BrOBr

CAS RN 18800-30-1

IUPAC Name: 1-bromo-4-(2-bromoethoxy)benzene
Synonyms: 1-bromo-4-(2-bromoethoxy)benzene
Molecular Weight (g/mol): 279.96
Molecular Formula: C8H8Br2O
InChi Key: SRGBSEMMGLSUMI-UHFFFAOYSA-N
SMILES: BrCCOC1=CC=C(Br)C=C1
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)