accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COOOHONH3CCH3CH3

CAS RN 18822-86-1

IUPAC Name: [3-carboxy-2-(octanoyloxy)propyl]trimethylazanium chloride
Synonyms: [3-carboxy-2-(octanoyloxy)propyl]trimethylazanium chloride
Molecular Weight (g/mol): 323.86
Molecular Formula: C15H30ClNO4
InChi Key: MYMFUYYXIJGKKU-UHFFFAOYNA-N
SMILES: [Cl-].CCCCCCCC(=O)OC(CC(O)=O)C[N+](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results