accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1883-78-9

H3CCH3SO

CAS RN 1883-78-9

IUPAC Name: 2-[(propan-2-ylsulfanyl)methyl]furan
Synonyms: 2-[(isopropylsulfanyl)methyl]furan
Molecular Weight (g/mol): 156.24
Molecular Formula: C8H12OS
InChi Key: WCHRNAKORAINOJ-UHFFFAOYSA-N
SMILES: CC(C)SCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results