CAS RN 1883429-22-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BI 9564
Potent and selective BRD9 and BRD7 inhibitor; orally active