CAS RN 189197-69-1
CAS RN 189197-69-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ OSC Inhibitor, Ro 48-8071
The OSC Inhibitor, Ro 48-8071, also referenced under CAS 189197-69-1, controls the biological activity of 2,3-oxidosqualene:lanosterol cyclase activity (OSC).
2
Tocris Bioscience™ Ro 48-8071 fumarate

2,3-Oxidosqualene cyclase (OSC) inhibitor; blocks cholesterol synthesis