Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 48-8071 fumarate
GREENER_CHOICESDP

Catalog No. 538950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-895-0 50 mg
53-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-895-0 Supplier Tocris Bioscience™ Supplier No. 5389/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

2,3-Oxidosqualene cyclase (OSC) inhibitor; blocks cholesterol synthesis

Ro 48-8071 fumarate is a 2,3-Oxidosqualene cyclase (OSC) inhibitor (IC50 = 6.5 nM); blocks cholesterol synthesis in HepG2 cells. Reduces viability of breast cancer cell lines. Also inhibits Ebola virus (EBOV) cell entry (IC50 = 1.74 μM).

Specifications

CAS 189197-69-1
Quantity 50 mg
Molecular Formula (4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone
Product Type Ro 48-8071 fumarate
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 564.44
Inhibitors Ro 48-8071 fumarate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.