CAS RN 1897379-34-8
CAS RN 1897379-34-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ SLM 6031434 hydrochloride
Selective sphingosine kinase 2 (Sphk2) inhibitor