CAS RN 192575-19-2
IUPAC Name:
tetrasodium 4-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]-2,5-dihydropyridin-2-ylidene}hydrazin-1-yl)benzene-1,3-disulfonate
Synonyms:
tetrasodium 4-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]pyridin-2-ylidene}hydrazin-1-yl)benzene-1,3-disulfonate
Molecular Weight (g/mol):
599.3
Molecular Formula:
C14H10N3Na4O12PS2
InChi Key:
ZNPHTQDEBMCNDG-UHFFFAOYSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].CC1=NC(=NNC2=C(C=C(C=C2)S([O-])(=O)=O)S([O-])(=O)=O)C(COP([O-])([O-])=O)=C(C=O)C1=O
1
–
1
of
1
results
1
R&D Systems™ PPADS tetrasodium salt
CAS: 192575-19-2 Molecular Formula: C14H10N3Na4O12PS2