Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PPADS tetrasodium salt
GREENER_CHOICESDP

Catalog No. 062550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-255-0 50 mg
06-251-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 06-255-0 Supplier R&D Systems™ Supplier No. 0625/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective P2 antagonist

PPADS tetrasodium salt is a non-selective P2 purinergic antagonist. Blocks recombinant P2X1, P2X2, P2X3, P2X5 (IC50 = 1 - 2.6 μM), native P2Y2-like (IC50 ∼ 0.9 mM), and recombinant P2Y4 (IC50 ∼ 15 mM) receptors. PPADS tetrasodium salt delays onset of calcium responses to mild hypoosmotic stress in cortical slices. PPADS tetrasodium salt blocks P2X7 and reduces clinical and histological graft-versus-host disease development in a humanised mouse model.

2'5'-isomer also available.

Chemical Identifiers

CAS 192575-19-2
Molecular Formula C14H10N3Na4O12PS2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 599.3
Purity >98%
Molecular Weight (g/mol) 599.296
Chemical Name or Material PPADS tetrasodium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.