CAS RN 193000-39-4
IUPAC Name:
5-{[7-(benzyloxy)quinazolin-4-yl]amino}-4-fluoro-2-methylphenol hydrochloride
Synonyms:
5-{[7-(benzyloxy)quinazolin-4-yl]amino}-4-fluoro-2-methylphenol hydrochloride
Molecular Weight (g/mol):
411.86
Molecular Formula:
C22H19ClFN3O2
InChi Key:
AVRHWGLIYGJSOD-UHFFFAOYSA-N
SMILES:
Cl.CC1=CC(F)=C(NC2=C3C=CC(OCC4=CC=CC=C4)=CC3=NC=N2)C=C1O
1
–
1
of
1
results
1
R&D Systems™ ZM 323881 hydrochloride
CAS: 193000-39-4 Molecular Formula: C22H19ClFN3O2