Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ZM 323881 hydrochloride
GREENER_CHOICESDP

Catalog No. 247510 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-751-0 10 mg
24-751 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-751-0 Supplier R&D Systems™ Supplier No. 2475/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective inhibitor of VEGFR-2

Potent and selective inhibitor of human vascular endothelial growth factor receptor 2 (VEGFR-2/KDR) activity. Selectively inhibits VEGFR-2 (IC50 = 2 nM) over VEGFR-1 and a range of other receptor tyrosine kinases such as PDGFRβ, FGFR1, EGFR and erbB2 (IC50 > 50 μM). Inhibits VEGF-A-induced endothelial cell proliferation in vitro (IC50 = 8 nM).

Chemical Identifiers

CAS 193000-39-4
Molecular Formula C22H19ClFN3O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 411.86
Purity >99%
Molecular Weight (g/mol) 411.861
Chemical Name or Material ZM 323881 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.