CAS RN 1947348-42-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BOP
Dual α9β1/α4β1 integrin inhibitor; preferentially mobilizes HSCs