CAS RN 1949837-12-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ARV 771
Potent BET bromodomain PROTAC™; also degrades BRD-tagged chimeric antigen receptors (CAR) in T cells