CAS RN 195055-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ R 121919 Hydrochloride
High affinity CRF1 antagonist; orally bioavailable