CAS RN 1950634-92-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ dBET6
Potent and selective (+)-JQ1 based Degrader (PROTAC™) targeting BET bromodomains, active in vivo