accessibility menu, dialog, popup

UserName

Viewing profile as user:
ICH3NH3COO

CAS RN 1952-15-4

IUPAC Name: 4-[(2,2-diphenylacetyl)oxy]-1,1-dimethylpiperidin-1-ium iodide
Synonyms: 4-[(2,2-diphenylacetyl)oxy]-1,1-dimethylpiperidin-1-ium iodide
Molecular Weight (g/mol): 451.35
Molecular Formula: C21H26INO2
InChi Key: WWJHRSCUAQPFQO-UHFFFAOYSA-M
SMILES: [I-].C[N+]1(C)CCC(CC1)OC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results