accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1953-02-2

H3CSHONHOHO

CAS RN 1953-02-2

IUPAC Name: 2-(2-sulfanylpropanamido)acetic acid
Synonyms: α mercaptopropionylglycine
Molecular Weight (g/mol): 163.19
Molecular Formula: C5H9NO3S
InChi Key: YTGJWQPHMWSCST-UHFFFAOYNA-N
SMILES: CC(S)C(=O)NCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)