accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1953-04-4

BrCH3ONH3C(S)(R)OH(S)OH

CAS RN 1953-04-4

IUPAC Name: hydrogen (1S,12S,14R)-9-methoxy-4-methyl-11-oxa-4-azatetracyclo[8.6.1.0¹,¹².0⁶,¹⁷]heptadeca-6,8,10(17),15-tetraen-14-ol bromide
Synonyms: hydrogen galantamine bromide
Molecular Weight (g/mol): 368.27
Molecular Formula: C17H22BrNO3
InChi Key: QORVDGQLPPAFRS-XPSHAMGMSA-N
SMILES: [H+].[Br-].COC1=CC=C2CN(C)CC[C@@]34C=C[C@H](O)C[C@@H]3OC1=C24
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)