accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19545-26-7

IUPAC Name: (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate
Synonyms: wortmannin wartmannin antibiotic SL-2052
Molecular Weight (g/mol): 428.44
Molecular Formula: C23H24O8
InChi Key: QDLHCMPXEPAAMD-QAIWCSMKSA-N
SMILES: COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)