Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wortmannin, 98%

Catalog No. AC328590050
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328590050 5 mg
AC328590010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328590050 Supplier Thermo Scientific Chemicals Supplier No. 328590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19545-26-7 | C23H24O8 | 428.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19545-26-7
Molecular Formula C23H24O8
Molecular Weight (g/mol) 428.44
MDL Number MFCD00133927
InChI Key QDLHCMPXEPAAMD-QAIWCSMKSA-N
Synonym wortmannin, wartmannin, antibiotic sl-2052, unii-xva4o219qw, xva4o219qw, pi 3-kinase inhibitor, 1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione, 1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate, 1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate, wortmanin
PubChem CID 312145
ChEBI CHEBI:52289
IUPAC Name (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate
SMILES COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O

Specifications

Melting Point 234.0°C to 240.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 mg
Beilstein 19, IV, 3134
Merck Index 15, 10251
Formula Weight 428.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Wortmannin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.