Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Wortmannin
GREENER_CHOICESDP

Catalog No. 12325 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-325 5 mg
12-321 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-325 Supplier R&D Systems™ Supplier No. 1232/5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, irreversible inhibitor of PI 3-kinase. Also inhibitor of PLK1

Potent, selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase) (IC50 = 2 - 4 nM). Also potently inhibits polo-like kinase 1 (PLK1) (IC50 = 5.8 nM).

Chemical Identifiers

CAS 19545-26-7
Molecular Formula C23H24O8

Specifications

Quantity 5 mg
Product Line Tocris
Formula Weight 428.44
Purity >98%
Molecular Weight (g/mol) 428.44
Chemical Name or Material Wortmannin
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.