CAS RN 1957203-01-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ YKL 5-124
Potent and selective CDK7 inhibitor; induces cell cycle arrest