accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClHClOOOONNNHNHNNH2CH3H3C

CAS RN 1962928-23-9

IUPAC Name: N-(2-aminoethyl)-2-[4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy]acetamide dihydrochloride
Synonyms: xanthine amine congener dihydrochloride
Molecular Weight (g/mol): 501.41
Molecular Formula: C21H30Cl2N6O4
InChi Key: KGNTWECSNGWFKE-UHFFFAOYSA-N
SMILES: Cl.Cl.CCCN1C2=C(NC(=N2)C2=CC=C(OCC(=O)NCCN)C=C2)C(=O)N(CCC)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results