accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHONHClOCH3

CAS RN 196868-63-0

IUPAC Name: 1-(5-chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea
Synonyms: 1-(5-chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea
Molecular Weight (g/mol): 341.8
Molecular Formula: C18H16ClN3O2
InChi Key: YBLWOZUPHDKFOT-UHFFFAOYSA-N
SMILES: COC1=C(NC(=O)NC2=C3C=CC=CC3=NC(C)=C2)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)