accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3SH

CAS RN 19788-49-9

IUPAC Name: ethyl 2-sulfanylpropanoate
Synonyms: ethyl 2-sulfanylpropanoate
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: LXXNWCFBZHKFPT-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)S
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results