CAS RN 1992047-61-6
IUPAC Name:
N-cyclopropyl-2-(propane-1-sulfonyl)-6-[1-(2,2,6,6-tetramethylpiperidin-4-yl)-1H-indol-5-yl]pyrimidin-4-amine
Synonyms:
N-cyclopropyl-2-(propane-1-sulfonyl)-6-[1-(2,2,6,6-tetramethylpiperidin-4-yl)indol-5-yl]pyrimidin-4-amine
Molecular Weight (g/mol):
495.69
Molecular Formula:
C27H37N5O2S
InChi Key:
XVECNLUKQDKOST-UHFFFAOYSA-N
SMILES:
CCCS(=O)(=O)C1=NC(=CC(NC2CC2)=N1)C1=CC=C2N(C=CC2=C1)C1CC(C)(C)NC(C)(C)C1
1
–
1
of
1
results
1
Tocris Bioscience™ NVS-CECR2-1, >98%
CAS: 1992047-61-6 Molecular Formula: C27H37N5O2S