Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVS-CECR2-1, >98%
GREENER_CHOICESDP

Catalog No. 574450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-445-0 50 mg
57-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-445-0 Supplier Tocris Bioscience™ Supplier No. 5744/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of cat eye syndrome chromosome region, candidate 2 (CECR2)

Potent and selective CECR2 (cat eye syndrome chromosome region, candidate 2) inhibitor (IC50 = 47 nM). Selective for CECR2 over a panel of bromodomains.

Chemical Identifiers

CAS 1992047-61-6
Molecular Formula C27H37N5O2S

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 495.68
Purity >98%
Molecular Weight (g/mol) 495.686
Chemical Name or Material N-Cyclopropyl-2-(propylsulfonyl)-6-(1-(2,2,6,6-tetramethylpiperidin-4-yl)-1H-indol-5-yl)pyrimidin-4-
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.