CAS RN 199522-35-5
IUPAC Name:
1-{3-[(5-bromopyridin-2-yl)[(3,4-dichlorophenyl)methyl]amino]propyl}-3-[3-(1H-imidazol-5-yl)propyl]thiourea
Synonyms:
1-{3-[(5-bromopyridin-2-yl)[(3,4-dichlorophenyl)methyl]amino]propyl}-3-[3-(3H-imidazol-4-yl)propyl]thiourea
Molecular Weight (g/mol):
556.35
Molecular Formula:
C22H25BrCl2N6S
InChi Key:
UREJDUPKGMFJRU-UHFFFAOYSA-N
SMILES:
ClC1=C(Cl)C=C(CN(CCCNC(=S)NCCCC2=CN=CN2)C2=NC=C(Br)C=C2)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ NNC 26-9100
CAS: 199522-35-5 Molecular Formula: C22H25BrCl2N6S