accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNNNNNHOCH3NHOOH

CAS RN 199986-75-9

IUPAC Name: 2-[(2-hydroxyethyl)(6-{[(4-methoxyphenyl)methyl]amino}-9-(propan-2-yl)-9H-purin-2-yl)amino]ethan-1-ol
Synonyms: 2-[(2-hydroxyethyl)(9-isopropyl-6-{[(4-methoxyphenyl)methyl]amino}purin-2-yl)amino]ethanol
Molecular Weight (g/mol): 400.48
Molecular Formula: C20H28N6O3
InChi Key: NQVIIUBWMBHLOZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC2=C3N=CN(C(C)C)C3=NC(=N2)N(CCO)CCO)C=C1
Form 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results