Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CVT 313
GREENER_CHOICESDP

Catalog No. 617450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-745-0 50 mg
61-741-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-745-0 Supplier R&D Systems™ Supplier No. 6174/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. cdk2 inhibitor

Cyclin-dependent kinase 2 (cdk2) inhibitor (reported IC50 values are 0.18 to 0.5 μM). Also inhibits cdk5 in sub-micromolar range (IC50 = 0.42 μM). Displays >7-fold selectivity for cdk2 and cdk5 over other cdks. Inhibits proliferation rat neonatal aortic smooth muscle cells and a range of tumor cells lines in vitro. Inhibits restenosis in a rat carotid artery injury model.

Chemical Identifiers

CAS 199986-75-9
Molecular Formula C20H28N6O3

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Product Line Tocris
Formula Weight 400.47
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 400.48
Chemical Name or Material 2,2'-[[6-[[(4-Methoxyphenyl)methyl]amino]-9-(1-methylethyl)-9H-purin-2-yl]imino]bis[ethanol]
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.