accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHCOOOHOOH(E/Z)

CAS RN 200189-97-5

IUPAC Name: but-2-enedioic acid; methyl({2-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-9-yl}methyl)(prop-2-yn-1-yl)amine
Synonyms: butenedioic acid; omigapil
Molecular Weight (g/mol): 391.42
Molecular Formula: C23H21NO5
InChi Key: SQAZQLMBEHYFJA-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(O)=O.CN(CC#C)CC1=CC2=CC=CC=C2OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)