Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CGP 3466B maleate
GREENER_CHOICESDP

Catalog No. 296650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-665-0 50 mg
29-661-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-665-0 Supplier R&D Systems™ Supplier No. 2966/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. GAPDH inhibitor; neuroprotective

CGP 3466B maleate is an orally active glyceraldehyde-3-phosphate dehydrogenase (GAPDH) inhibitor. Exhibits antiapoptotic activity via inhibition of GAPDH nuclear translocation and mitochondrial complex 1-dependent H2O2 release. CGP 3466B inhibits colon cancer proliferation in vitro and displays neuroprotective activity in vivo.

Chemical Identifiers

CAS 200189-97-5
Molecular Formula C23H21NO5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 391.42
Purity >99%
Molecular Weight (g/mol) 391.423
Chemical Name or Material CGP 3466B maleate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.