accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NClNNOHOOHOHN

CAS RN 2004-07-1

IUPAC Name: 2-(2-amino-6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-(2-amino-6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 301.69
Molecular Formula: C10H12ClN5O4
InChi Key: TXWHPSZYRUHEGT-UHFFFAOYNA-N
SMILES: NC1=NC(Cl)=C2N=CN(C3OC(CO)C(O)C3O)C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results