accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2NOOHOHO

CAS RN 201730-11-2

IUPAC Name: 4-[amino(carboxy)methyl]benzene-1,2-dicarboxylic acid
Synonyms: DCPG
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: IJVMOGKBEVRBPP-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC(C(O)=O)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)