accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 20184-91-2

IUPAC Name: non-4-yne
Synonyms: 4-nonyne
Molecular Weight (g/mol): 124.23
Molecular Formula: C9H16
InChi Key: RAYTXPDVKZFUEI-UHFFFAOYSA-N
SMILES: CCCCC#CCCC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)