accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOH2NOOH

CAS RN 201943-63-7

IUPAC Name: 2-[1-amino-1-carboxy-2-(9H-xanthen-9-yl)ethyl]cyclopropane-1-carboxylic acid
Synonyms: 2-[1-amino-1-carboxy-2-(9H-xanthen-9-yl)ethyl]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 353.37
Molecular Formula: C20H19NO5
InChi Key: VLZBRVJVCCNPRJ-UHFFFAOYNA-N
SMILES: NC(CC1C2=CC=CC=C2OC2=CC=CC=C12)(C1CC1C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results