Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LY 341495
GREENER_CHOICESDP

Catalog No. 120910 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-091-0 10 mg
12-091 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-091-0 Supplier R&D Systems™ Supplier No. 1209/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent and selective group II mGlu antagonist

Highly potent and selective group II metabotropic glutamate receptor antagonist (Ki/IC50 values are 2.3, 1.3, 173, 990, 6800, 8200 and 22000 nM for human mGlu2, mGlu3, mGlu8, mGlu7a, mGlu1a, mGlu5a and mGlu4a receptors respectively). Readily brain penetrant and active in vivo.

Disodium salt (Cat. No. 4062) also available.

Chemical Identifiers

CAS 201943-63-7
Molecular Formula C20H19NO5

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 353.37
Purity >98%
Molecular Weight (g/mol) 353.374
Chemical Name or Material LY 341495
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.