accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20217-01-0

BrOOBr

CAS RN 20217-01-0

IUPAC Name: 2-[(2,4-dibromophenoxy)methyl]oxirane
Synonyms: 2-(2,4-dibromophenoxymethyl)oxirane
Molecular Weight (g/mol): 307.97
Molecular Formula: C9H8Br2O2
InChi Key: NFWLWLQSZIJYFR-UHFFFAOYNA-N
SMILES: BrC1=CC=C(OCC2CO2)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)