CAS RN 2022-85-7
5-Fluorocytosine 98.0+%, TCI America™
CAS: 2022-85-7 Molecular Formula: C4H4FN3O Molecular Weight (g/mol): 129.09 MDL Number: MFCD00006035,MFCD00179326,MFCD03547958 InChI Key: XRECTZIEBJDKEO-UHFFFAOYSA-N Synonym: 5-fluorocytosine,flucytosine,ancotil,ancobon,flucytosin,5-fluorocystosine,fluocytosine,5-fc,5-fluorocytosin,fluorcytosine PubChem CID: 3366 ChEBI: CHEBI:5100 IUPAC Name: 6-amino-5-fluoro-1,2-dihydropyrimidin-2-one SMILES: NC1=C(F)C=NC(=O)N1
5-Fluorocytosine, 99+%
CAS: 2022-85-7 Molecular Formula: C4H4FN3O Molecular Weight (g/mol): 129.09 MDL Number: MFCD00006035,MFCD00179326,MFCD03547958 InChI Key: XRECTZIEBJDKEO-UHFFFAOYSA-N Synonym: 5-fluorocytosine,flucytosine,ancotil,ancobon,flucytosin,5-fluorocystosine,fluocytosine,5-fc,5-fluorocytosin,fluorcytosine PubChem CID: 3366 ChEBI: CHEBI:5100 IUPAC Name: 6-amino-5-fluoro-1H-pyrimidin-2-one SMILES: NC1=C(F)C=NC(=O)N1
Flucytosine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Flucytosine is an antimycotic drug that can be effectively utilized against severe Candida and Cryptococcus infections.
5-Fluorocytosine, 98+%
CAS: 2022-85-7 Molecular Formula: C4H4FN3O Molecular Weight (g/mol): 129.09 MDL Number: MFCD00006035,MFCD00179326,MFCD03547958 InChI Key: XRECTZIEBJDKEO-UHFFFAOYSA-N Synonym: 5-fluorocytosine,flucytosine,ancotil,ancobon,flucytosin,5-fluorocystosine,fluocytosine,5-fc,5-fluorocytosin,fluorcytosine PubChem CID: 3366 ChEBI: CHEBI:5100 SMILES: NC1=C(F)C=NC(=O)N1
5-Fluorocytosine, 98%, Thermo Scientific™
CAS: 2022-85-7 Molecular Formula: C4H4FN3O Molecular Weight (g/mol): 129.09 MDL Number: MFCD00006035,MFCD00179326,MFCD03547958 InChI Key: XRECTZIEBJDKEO-UHFFFAOYSA-N Synonym: 5-fluorocytosine,flucytosine,ancotil,ancobon,flucytosin,5-fluorocystosine,fluocytosine,5-fc,5-fluorocytosin,fluorcytosine PubChem CID: 3366 ChEBI: CHEBI:5100 IUPAC Name: 6-amino-5-fluoro-1,2-dihydropyrimidin-2-one SMILES: NC1=C(F)C=NC(=O)N1