Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorocytosine, 98+%

Catalog No. AAL16496MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL16496MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL16496MD Supplier Thermo Scientific Chemicals Supplier No. L16496MD
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2022-85-7 | C4H4FN3O | 129.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
5-Fluorocytosine acts as an antidiabetic, antifungal and antimicrobial agent. It is useful for the treatment of serious infections arises due to susceptible strains of Candida or Cryptococcus neoformans and chromomycosis. Further, it is employed in studies on TMP biosynthesis.

Solubility
Soluble in water and hydrochloric acid.

Notes
Light Sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
SMILES NC1=C(F)C=NC(=O)N1

Specifications

Melting Point ∽297°C (decomposition)
Quantity 250 mg
Beilstein 127285
Sensitivity Light sensitive
Merck Index 14,4125
Solubility Information Soluble in water and hydrochloric acid.
IUPAC Name 6-amino-5-fluoro-1H-pyrimidin-2-one
Formula Weight 129.09
Percent Purity ≥98%
Chemical Name or Material 5-Fluorocytosine
Hazard Category Warning
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-968-7
RTECSNumber HA6040000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.