Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorocytosine, 98%, Thermo Scientific™

Catalog No. AC327962500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327962500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC327962500 Supplier Thermo Scientific Chemicals Supplier No. 327962500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
IUPAC Name 6-amino-5-fluoro-1,2-dihydropyrimidin-2-one
SMILES NC1=C(F)C=NC(=O)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 250 mg
Beilstein 25, 3688
Merck Index 15, 4156
Formula Weight 129.09
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorocytosine, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.