CAS RN 202645-74-7
IUPAC Name:
3-[bis(4-fluorophenyl)methoxy]-8-butyl-8-azabicyclo[3.2.1]octane hydrochloride
Synonyms:
3-[bis(4-fluorophenyl)methoxy]-8-butyl-8-azabicyclo[3.2.1]octane hydrochloride
Molecular Weight (g/mol):
421.96
Molecular Formula:
C24H30ClF2NO
InChi Key:
RHYMGBAYGRUKNZ-UHFFFAOYNA-N
SMILES:
Cl.CCCCN1C2CCC1CC(C2)OC(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
R&D Systems™ JHW 007 hydrochloride
CAS: 202645-74-7 Molecular Formula: C24H30ClF2NO