CAS RN 202821-81-6
IUPAC Name:
3-[(5-chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-2,3-dihydro-1,3,4-oxadiazol-2-one
Synonyms:
3-[(5-chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2-one
Molecular Weight (g/mol):
370.71
Molecular Formula:
C16H10ClF3N2O3
InChi Key:
QKOWACXSXTXRKA-UHFFFAOYSA-N
SMILES:
OC1=C(CN2N=C(OC2=O)C2=CC=C(C=C2)C(F)(F)F)C=C(Cl)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ BMS 191011
CAS: 202821-81-6 Molecular Formula: C16H10ClF3N2O3