accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202925-04-0

BrClFBr

CAS RN 202925-04-0

IUPAC Name: 1,5-dibromo-2-chloro-3-fluorobenzene
Synonyms: 1,5-dibromo-2-chloro-3-fluorobenzene
Molecular Weight (g/mol): 288.34
Molecular Formula: C6H2Br2ClF
InChi Key: WRJXIIFHUQBLSW-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(Br)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results