accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHSOOHOSOOOHSOOOHNHONHONHONHONHSOOOHSOOOHSOOOHNaNaNaNaNaNa

CAS RN 202983-32-2

IUPAC Name: 8-{4-[4-({[4-({4-[(4,6,8-trisulfonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}naphthalene-1,3,5-trisulfonic acid hexasodium
Synonyms: 8-{4-[4-({[4-({4-[(4,6,8-trisulfonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}naphthalene-1,3,5-trisulfonic acid hexasodium
Molecular Weight (g/mol): 1407.14
Molecular Formula: C49H36N6Na6O23S6
InChi Key: RRKORSRLXJQAGK-UHFFFAOYSA-N
SMILES: [Na].[Na].[Na].[Na].[Na].[Na].OS(=O)(=O)C1=CC(=C2C(NC(=O)C3=CC=C(NC(=O)C4=CC=C(NC(=O)NC5=CC=C(C=C5)C(=O)NC5=CC=C(C=C5)C(=O)NC5=CC=C(C6=CC(=CC(=C56)S(O)(=O)=O)S(O)(=O)=O)S(O)(=O)=O)C=C4)C=C3)=CC=C(C2=C1)S(O)(=O)=O)S(O)(=O)=O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results