CAS RN 2036081-86-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ JNJ 55511118
Selective, high affinity negative modulator of AMPA receptors containing TARP-γ8