CAS RN 2037-26-5
Toluene-d8, for NMR, 99+ atom % D
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 MDL Number: MFCD00044638 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, 99.6%(Isotopic), Thermo Scientific Chemicals
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 MDL Number: MFCD00044638 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, for NMR, 99.5 atom % D, with 0.03% TMS
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 MDL Number: MFCD00044638 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, for NMR, 99.5+ atom % D
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 MDL Number: MFCD00044638 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N IUPAC Name: 1-(²H₃)methyl(²H₅)benzene SMILES: [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Toluene-d8, for NMR, 100.0 atom % D, packaged in 0.50 ml ampoules
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 MDL Number: MFCD00044638 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, for NMR, 99.5 atom % D, AcroSeal™
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d{8}, 100% (Isotopic), Thermo Scientific Chemicals
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 MDL Number: MFCD00044638 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, for NMR, 99.8 atom % D
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N IUPAC Name: 1-(²H₃)methyl(²H₅)benzene SMILES: [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Toluene-d8, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N IUPAC Name: 1-(²H₃)methyl(²H₅)benzene SMILES: [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Toluene-d8, for NMR, 100.0 atom % D
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8 Solution, 2000μg/mL in methanol, Ultra Scientific
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, Thermo Scientific™
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N Synonym: toluene-d8,2h8 toluene,perdeuteriotoluene,benzene-d5, methyl-d3,perdeuterotoluene,benzene-d5-, methyl-d3,1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene,toluene d8,toluene-d8, 99 atom % d,toluene-d8, 99.6 atom % d PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1
Toluene-d8, SPEX CertiPrep™
CAS: 2037-26-5 Molecular Formula: C7H8 Molecular Weight (g/mol): 100.19 InChI Key: YXFVVABEGXRONW-JGUCLWPXSA-N PubChem CID: 74861 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene SMILES: CC1=CC=CC=C1